Assay Details
| Gene Symbol: A630095N17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243090.1 | NP_001230019.1 | 2 - 3 | 250 | 157 | |
| NM_001243091.1 | NP_001230020.1 | 2 - 3 | 163 | 157 | ||
| GenBank mRNA | AK080399.1 | - | 2 - 3 | 164 | 157 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
tubulin, alpha 4A |
Tuba4a |
M[a]4, Tuba4 |
-235.0 | Sense | Chr. 1 - 75214974 - 75219860 on NCBI Build GRCm38 |
| Downstream |
DnaJ (Hsp40) homolog, subfamily B, member 2 |
Dnajb2 |
2700059H22Rik, Dnajb10, Hsj1, mDj8 |
13.0kb | Anti-sense | Chr. 1 - 75236403 - 75245692 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

A630095N17Rik
Genomic Map