Assay Details
| Gene Symbol: Prkaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178143.2 | NP_835279.2 | 5 - 6 | 670 | 68 | |
| GenBank mRNA | AK044030.1 | - | 5 - 6 | 506 | 68 | |
| AK134573.1 | - | 5 - 6 | 528 | 68 | ||
| BC138565.1 | - | 5 - 6 | 679 | 8861585 | 68 | |
| BC138566.1 | - | 5 - 6 | 679 | 8861586 | 68 | |
| BY718361.1 | - | 5 - 6 | 525 | 68 | ||
| FJ588504.1 | - | 4 - 5 | 428 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prkaa2
Genomic Map