Assay Details
| Gene Symbol: 2010010A06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045393.1 | 2 - 3 | 305 | 61 | ||
| GenBank mRNA | AK019891.1 | - | 2 - 3 | 306 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

2010010A06Rik
Genomic Map