Assay Details
Gene Symbol: Gm19519 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK036854.1 | - | 2 - 3 | 612 | 124 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700011I03 gene | 1700011I03Rik | 4930583I06Rik | -47.3kb | Anti-sense | Chr. 18 - 57533798 - 57731067 on NCBI Build GRCm38 |
Downstream |
solute carrier family 12, member 2 |
Slc12a2 |
9330166H04Rik, Nkcc1, mBSC2, sy-ns |
99.2kb | Anti-sense | Chr. 18 - 57878513 - 57946821 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631670 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105104 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13812869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13765359 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738281 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738141 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738283 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738142 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13964580 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738218 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13598958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738210 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738290 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738248 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738259 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738241 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738230 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738209 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13882036 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738203 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13821396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13964578 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13606409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13703245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738226 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738171 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13606410 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738246 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738199 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738206 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738265 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13703260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738214 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738137 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13786619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13812853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738140 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13703274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31738208 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 634066 | - | - |
