Assay Details
| Gene Symbol: 4930546C10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_038051.1 | 2 - 3 | 1674 | 73 | ||
| GenBank mRNA | AK019744.1 | - | 2 - 3 | 1675 | 73 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6356 | Mir6356 |
Gm27461, mmu-mir-6356 |
-205.1kb | Anti-sense | Chr. 18 - 68739400 - 68739501 on NCBI Build GRCm38 |
| Downstream | microRNA 145b | Mir145b |
Gm27644, mmu-mir-145b |
74.7kb | Sense | Chr. 18 - 69022199 - 69022273 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4930546C10Rik
Genomic Map