Assay Details
Gene Symbol: Arhgef16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112744.1 | NP_001106215.1 | 9 - 10 | 1488 | 57 | |
GenBank mRNA | AK042195.1 | - | 9 - 10 | 1484 | 57 | |
AK153708.1 | - | 9 - 10 | 1649 | 57 | ||
BC020030.1 | - | 6 - 7 | 649 | 57 | ||
BC026661.1 | - | 1 - 2 | 501 | 57 | ||
BC044805.1 | - | 8 - 9 | 2902 | 57 | ||
BC095966.1 | - | 7 - 8 | 785 | 57 | ||
BC137847.1 | - | 9 - 10 | 1529 | 8860866 | 57 | |
BC144970.1 | - | 9 - 10 | 1529 | 9053518 | 57 | |
BC144971.1 | - | 9 - 10 | 1552 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
