Assay Details
| Gene Symbol: Arhgef16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112744.1 | NP_001106215.1 | 12 - 13 | 1911 | 74 | |
| GenBank mRNA | AK042195.1 | - | 12 - 13 | 1907 | 74 | |
| AK153708.1 | - | 12 - 13 | 2072 | 74 | ||
| BC020030.1 | - | 9 - 10 | 1072 | 74 | ||
| BC044805.1 | - | 11 - 12 | 3325 | 74 | ||
| BC095966.1 | - | 10 - 11 | 1208 | 74 | ||
| BC137847.1 | - | 12 - 13 | 1952 | 8860866 | 74 | |
| BC144970.1 | - | 12 - 13 | 1952 | 9053518 | 74 | |
| BC144971.1 | - | 12 - 13 | 1975 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef16
Genomic Map