Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293685.1 | NP_001280614.1 | 4 - 5 | 740 | 87 | |
| GenBank mRNA | BC089321.1 | - | 4 - 5 | 447 | 6839437 | 87 |
More Information
Set Membership: |
|

Vti1a
Genomic Map