Assay Details
| Gene Symbol: Vti1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293685.1 | NP_001280614.1 | 5 - 6 | 761 | 73 | |
| GenBank mRNA | BC089321.1 | - | 5 - 6 | 468 | 6839437 | 73 |
More Information
Set Membership: |
|

Vti1a
Genomic Map