Assay Details
Gene Symbol: Riok3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024182.4 | NP_077144.2 | 3 - 4 | 393 | 91 | |
GenBank mRNA | AK004748.1 | - | 3 - 4 | 485 | 91 | |
AK131635.1 | - | 3 - 4 | 397 | 91 | ||
AK134327.1 | - | 3 - 4 | 485 | 91 | ||
AK147074.1 | - | 3 - 4 | 394 | 91 | ||
AK167309.1 | - | 3 - 4 | 489 | 91 | ||
AK170882.1 | - | 3 - 4 | 473 | 91 | ||
BC033271.1 | - | 3 - 4 | 431 | 4016197 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
