Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100004136.mm.1 |
NR_045914.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004136.mm.1 |
NR_045914.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004136.mm.1 |
TC0100000517.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100004139.mm.1 |
NR_045914.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004139.mm.1 |
NR_045914.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004139.mm.1 |
NR_045915.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004139.mm.1 |
NR_045915.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100004139.mm.1 |
TC0100000517.mm.1 |
MTA TC |