Assay Details
Gene Symbol: Gm553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC051519.1 | - | 474 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2132059 | BC051519.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 848743 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 913818 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 252221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441280 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 734844 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909320 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102552 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 921390 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441632 | AK003745.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441632 | AK156823.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441632 | BC038330.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 356160 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858031 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519284 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | AK049237.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | AK053860.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | AK147246.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | AK147269.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | AK220182.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | BC017516.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 349550 | NM_146001.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236256 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72930 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893863 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184778 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616496 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180226 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169452 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81974 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65249 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155249 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P131437 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2132059 | BC051519.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04097147 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P131437 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04097098 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04097122 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111130 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04097147 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P131437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01053542 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227707 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227974 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227669 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227694 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227763 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00491272 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227794 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227802 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00023230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227818 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01053554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227790 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227788 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01073740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227808 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227792 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227676 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227681 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227762 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00951231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00162447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227753 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227671 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227995 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227813 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227665 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227663 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00141132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227815 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20227705 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105570563 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105570563 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1218486 | - | - |
mouse-6 v1.1 (Illumina) | GI_38049483-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105570563 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105570563 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1218486 | - | - |
