Assay Details
| Gene Symbol: Carf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285463.1 | NP_001272392.1 | 14 - 15 | 1763 | 86 | |
| NM_001285473.1 | NP_001272402.1 | 10 - 11 | 1353 | 86 | ||
| NM_139150.5 | NP_631889.1 | 12 - 13 | 1636 | 86 | ||
| NR_104344.1 | 8 - 9 | 1088 | 86 | |||
| GenBank mRNA | AB221600.1 | - | 11 - 12 | 1419 | 86 | |
| AF454947.1 | - | 12 - 13 | 1605 | 86 | ||
| AK044738.1 | - | 14 - 15 | 1764 | 86 | ||
| BC138644.1 | - | 11 - 12 | 1542 | 8861664 | 86 | |
| BC138645.1 | - | 11 - 12 | 1542 | 8861665 | 86 | |
| EU170502.1 | - | 10 - 11 | 1334 | 86 | ||
| EU170503.1 | - | 8 - 9 | 1069 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Carf
Genomic Map