Assay Details
Gene Symbol: 4933424G05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045372.1 | 2 - 3 | 190 | 73 | ||
GenBank mRNA | AK016892.1 | - | 2 - 3 | 191 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | potassium channel tetramerisation domain containing 1 | Kctd1 |
4933402K10Rik, AI661543, AW553000 |
Anti-sense | Chr. 18 - 14968685 - 15151446 on NCBI Build GRCm38 | |
Downstream |
aldehyde dehydrogenase 9, subfamily A1 pseudogene |
Gm2629 | 223.5kb | Sense | Chr. 18 - 15213100 - 15214264 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
