Assay Details
Gene Symbol: 4933406J09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016696.1 | - | 6 - 7 | 1111 | 149 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ets variant 6 | Etv6 |
AW123102, AW557856, Tel |
-46.1kb | Anti-sense | Chr. 6 - 134034583 - 134270158 on NCBI Build GRCm38 |
Downstream | BCL2-like 14 (apoptosis facilitator) | Bcl2l14 |
4930452K23Rik, 4933405K19Rik, 9030625M01Rik, AI429211, Bclg |
78.1kb | Anti-sense | Chr. 6 - 134396329 - 134438741 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668249 | AK005770.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668249 | AK078306.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668249 | BC099973.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668249 | NM_001081347.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 85214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145677 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 362822 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 782849 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945913 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 922687 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625855 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621065 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 622432 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 574262 | AK089860.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360473 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871673 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713962 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 91066 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 851503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 443400 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963910 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 295512 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 376337 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450164 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 926534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385536 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907183 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 403063 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 407447 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861394 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 706275 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865702 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9745 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170264 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 145429 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 132938 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92474 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 86409 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72548 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151019 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06075775 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06075766 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06075876 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151019 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01449064 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06075805 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128749 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01449064 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06075805 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861291 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05338186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861249 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05311839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05603045 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861245 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861243 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05176516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05603034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861197 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05603040 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05311836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861298 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05816069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861303 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05061568 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05111317 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861241 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05816066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05111315 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861308 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861293 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861266 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05772969 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861208 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05753035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05603043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05626156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05243828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05626158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05868304 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05648386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861239 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861237 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05373849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05245399 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861268 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05463624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24861252 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1431520_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431520_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1431520_at | - | - |
Mouse Expression Set 430 | 1431520_at | - | - |
Mouse Expression Set 430 | 1431520_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431520_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431520_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 385682 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101780050 | - | - |
