Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400007320.mm.1 |
NR_045887.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007320.mm.1 |
NR_045887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007320.mm.1 |
TC0400001069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007318.mm.1 |
NR_045887.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007318.mm.1 |
NR_045887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007318.mm.1 |
TC0400001069.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007319.mm.1 |
NR_045887.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007319.mm.1 |
NR_045887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007319.mm.1 |
TC0400001069.mm.1 |
MTA TC |