Assay Details
| Gene Symbol: Sos2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135559.1 | NP_001129031.1 | 5 - 6 | 965 | 160 | |
| GenBank mRNA | Z11664.1 | - | 4 - 5 | 609 | 160 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sos2
Genomic Map