Assay Details
Gene Symbol: 3110002H16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029623.2 | NP_083899.1 | 19 - 20 | 1969 | 94 | |
GenBank mRNA | AF003352.1 | - | 7 - 8 | 855 | 94 | |
AK013979.1 | - | 13 - 14 | 1355 | 94 | ||
AK144623.1 | - | 19 - 20 | 1970 | 94 | ||
AK154240.1 | - | 19 - 20 | 1964 | 94 | ||
AK159691.1 | - | 19 - 20 | 1964 | 94 | ||
BC012285.1 | - | 18 - 19 | 2132 | 94 | ||
BC021846.1 | - | 19 - 20 | 1952 | 5027035 | 94 | |
BC044861.1 | - | 4 - 5 | 3610 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499179 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499176 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499177 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499178 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124445 | BC044861.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124445 | BC012285.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124445 | BC021846.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124445 | AK154240.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P124445 | AK159691.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320013 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 812091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 141530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 260640 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406478 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517648 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 806093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 689786 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687813 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 439949 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 359326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 121210 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168561 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117498 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170439 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124445 | BC044861.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124445 | BC012285.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124445 | BC021846.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124445 | AK154240.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P124445 | AK159691.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499192 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499160 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499166 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499158 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499157 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499171 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499167 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499175 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499156 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499181 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499168 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499163 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31499187 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08913934 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170439 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08913880 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117498 | - | - |
Whole Mouse Genome G4122A | A_51_P370101 | BC021846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P112931 | BC012285.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P112931 | BC021846.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P112931 | AK154240.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P112931 | AK159691.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P112931 | AK013979.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124445 | BC044861.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124445 | BC012285.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124445 | BC021846.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124445 | AK154240.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124445 | AK159691.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117498 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170439 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13916409 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13916407 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13916415 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499179 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499178 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499177 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31499176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499190 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499272 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499250 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499288 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499239 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499260 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499303 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499238 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499313 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499294 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499324 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499308 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499320 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31499248 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13916409 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499201 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13916416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499223 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499216 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13790505 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499153 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13773551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13828105 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499211 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13828108 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13874061 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499219 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499224 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13652821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499166 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499221 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499207 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13874063 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13783253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499168 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13603176 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499175 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499204 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13680642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499196 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499161 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31499172 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10453920 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453921 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453928 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453932 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453927 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453936 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453923 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453924 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453937 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453926 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453938 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453931 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453934 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453929 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453919 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453933 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453930 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453922 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453935 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10453925 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431330_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456716_PM_s_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10453918 | - | - |
Mouse Genome 430 2.0 Array | 1456716_PM_s_at | - | - |
Mouse Genome 430 2.0 Array | 1456716_s_at | - | - |
Mouse Genome 430 2.0 Array | 1431330_at | - | - |
Mouse Genome 430 2.0 Array | 1431330_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76482_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76482_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800000706.mm.1 | NM_029623.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800000706.mm.1 | NM_029623.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800000706.mm.1 | TC1800000100.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800000705.mm.1 | NM_029623.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800000705.mm.1 | NM_029623.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800000705.mm.1 | TC1800000100.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431330_PM_at | - | - |
Mouse Expression Set 430 | 1431330_at | - | - |
Mouse Expression Set 430 | 1456716_s_at | - | - |
Mouse Expression Set 430 | 1456716_PM_s_at | - | - |
Murine Genome U74v2 Set | 160433_PM_at | - | - |
Murine Genome U74v2 Set | 160433_at | - | - |
Mouse Genome 430A 2.0 Array | 1456716_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1431330_at | - | - |
Mouse Genome 430A 2.0 Array | 1431330_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1456716_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456716_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456716_PM_s_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431330_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1431330_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1570717 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1570717 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3940670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003940670 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001570717 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2747330 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2747330 | - | - |
mouse-6 v1.1 (Illumina) | scl076482.18_130-S | - | - |
mouse-6 v1.1 (Illumina) | scl00001.1_0-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1570717 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3940670 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl076482.18_130-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00001.1_0-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940670 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570717 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2747331 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1250418 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2747331 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl076482.18_130-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl00001.1_0-S | - | - |
