Assay Details
| Gene Symbol: Npc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008720.2 | NP_032746.2 | 14 - 15 | 2395 | 76 | |
| GenBank mRNA | AF003348.1 | - | 14 - 15 | 2372 | 76 | |
| AK154282.1 | - | 14 - 15 | 2315 | 76 | ||
| AK155379.1 | - | 14 - 15 | 2315 | 76 | ||
| BC052437.1 | - | 14 - 15 | 2395 | 6405214 | 76 | |
| BC054539.1 | - | 14 - 15 | 2395 | 6405214 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Npc1
Genomic Map