Assay Details
Gene Symbol: 0610025J13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002663.1 | - | 1 - 2 | 57 | 65 | |
AK021025.1 | - | 1 - 2 | 107 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear factor I/A | Nfia |
1110047K16Rik, 9430022M17Rik, CTF, NF1-A, NF1A |
-202.5kb | Sense | Chr. 4 - 97581717 - 98118876 on NCBI Build GRCm38 |
Downstream | TM2 domain containing 1 | Tm2d1 |
2310026L18Rik, AA990549, Bbp |
32.5kb | Anti-sense | Chr. 4 - 98355370 - 98383281 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241413 | AK021025.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P241413 | AK021025.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546611 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491188 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 270704 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109717 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P114454 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P474699 | AK002663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P474699 | AK002663.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241413 | AK021025.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P241413 | AK021025.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109717 | - | - |
Whole Mouse Genome G4122A | A_52_P474699 | AK002663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P474699 | AK002663.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241413 | AK021025.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P241413 | AK021025.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03451340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137905 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03759132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03548933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03759139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137972 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137866 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03799145 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137854 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137996 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137863 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377596 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137926 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137851 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137837 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137867 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03824489 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03301783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23138004 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137897 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137852 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137911 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137919 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137845 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137859 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137934 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137861 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03563564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23138001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03718398 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03288950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03288946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137869 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137839 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03608367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03260498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03548929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03322895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137954 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137856 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137939 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03401539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137858 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03548940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137872 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137865 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137933 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03354638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137921 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03548928 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03377597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137874 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03288951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137961 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137885 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137944 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137932 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23137864 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10506057 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432532_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432532_at | - | - |
Mouse Genome 430 2.0 Array | 1432532_PM_at | - | - |
Mouse Expression Set 430 | 1432532_at | - | - |
Mouse Expression Set 430 | 1432532_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432532_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432532_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 782000 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105690520 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1237506 | - | - |
