Assay Details
Gene Symbol: 0610025J13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK002663.1 | - | 2 - 3 | 238 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear factor I/A | Nfia |
1110047K16Rik, 9430022M17Rik, CTF, NF1-A, NF1A |
-203.4kb | Sense | Chr. 4 - 97581717 - 98118876 on NCBI Build GRCm38 |
Downstream | TM2 domain containing 1 | Tm2d1 |
2310026L18Rik, AA990549, Bbp |
23.6kb | Anti-sense | Chr. 4 - 98355370 - 98383281 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
