Assay Details
| Gene Symbol: Dnajc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164583.1 | NP_001158055.1 | 2 - 3 | 410 | 75 | |
| NM_001164584.1 | NP_001158056.1 | 2 - 3 | 355 | 75 | ||
| NM_001164585.1 | NP_001158057.1 | 3 - 4 | 420 | 75 | ||
| NM_198412.2 | NP_940804.1 | 3 - 4 | 329 | 75 | ||
| GenBank mRNA | AK122293.1 | - | 2 - 3 | 331 | 75 | |
| AK147570.1 | - | 3 - 4 | 292 | 75 | ||
| AK147637.1 | - | 3 - 4 | 292 | 75 | ||
| AK163657.1 | - | 2 - 3 | 229 | 75 | ||
| BC060734.1 | - | 2 - 3 | 410 | 6409942 | 75 | |
| BC064460.1 | - | 3 - 4 | 298 | 6821822 | 75 | |
| CJ111757.1 | - | 2 - 3 | 248 | 75 | ||
More Information
Set Membership: |
|

Dnajc6
Genomic Map