Assay Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286181.1 | NP_001273110.1 | 6 - 7 | 1236 | 95 | |
| NM_001286182.1 | NP_001273111.1 | 6 - 7 | 1193 | 95 | ||
| NM_010728.3 | NP_034858.2 | 5 - 6 | 1613 | 95 | ||
| GenBank mRNA | AK135213.1 | - | 5 - 6 | 744 | 95 | |
| AK141995.1 | - | 5 - 6 | 1499 | 95 | ||
| AK159199.1 | - | 5 - 6 | 1416 | 95 | ||
| AK159241.1 | - | 5 - 6 | 1392 | 95 | ||
| AK159263.1 | - | 5 - 6 | 1453 | 95 | ||
| AK159368.1 | - | 5 - 6 | 1375 | 95 | ||
| AK159682.1 | - | 5 - 6 | 1326 | 95 | ||
| AK159796.1 | - | 5 - 6 | 1387 | 95 | ||
| AK160096.1 | - | 5 - 6 | 1387 | 95 | ||
| AK164634.1 | - | 6 - 7 | 1194 | 95 | ||
| AK166279.1 | - | 5 - 6 | 1321 | 95 | ||
| AK166386.1 | - | 1457 | 95 | |||
| AK166442.1 | - | 1613 | 95 | |||
| BC018439.1 | - | 5 - 6 | 1464 | 2655752 | 95 | |
| M65142.1 | - | 5 - 6 | 1381 | 95 | ||
| M65143.1 | - | 2 - 3 | 240 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lox
Genomic Map