Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300007565.mm.1 |
NM_001102407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007565.mm.1 |
NM_001102407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007565.mm.1 |
NM_025875.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007565.mm.1 |
NM_025875.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007565.mm.1 |
TC0300000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007564.mm.1 |
NM_001102407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007564.mm.1 |
NM_001102407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007564.mm.1 |
NM_025875.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007564.mm.1 |
NM_025875.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007564.mm.1 |
TC0300000953.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300007563.mm.1 |
NM_001102407.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007563.mm.1 |
NM_001102407.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007563.mm.1 |
NM_025875.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007563.mm.1 |
NM_025875.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300007563.mm.1 |
TC0300000953.mm.1 |
MTA TC |