Assay Details
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027126.4 | NP_081402.3 | 2 - 3 | 320 | 74 | |
GenBank mRNA | AJ557515.1 | - | 2 - 3 | 320 | 74 | |
AK014082.1 | - | 2 - 3 | 292 | 74 | ||
BC022603.1 | - | 2 - 3 | 344 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK014082.1, BC022603.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
