Assay Details
| Gene Symbol: Polr3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028925.1 | NP_083201.1 | 12 - 13 | 1423 | 64 | |
| GenBank mRNA | AK016716.1 | - | 12 - 13 | 1450 | 64 | |
| BC147485.1 | - | 12 - 13 | 1423 | 9056238 | 64 | |
| BC151028.1 | - | 12 - 13 | 1423 | 9087941 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr3c
Genomic Map