Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700025579.mm.1 |
TC0700002857.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025579.mm.1 |
NM_178704.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025579.mm.1 |
NM_178704.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025580.mm.1 |
TC0700002857.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025580.mm.1 |
NM_178704.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025580.mm.1 |
NM_178704.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025577.mm.1 |
TC0700002857.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025577.mm.1 |
NM_178704.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025577.mm.1 |
NM_178704.3 |
Refseq |