Assay Details
| Gene Symbol: Dpy19l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178704.3 | NP_848819.2 | 7 - 8 | 963 | 86 | |
| GenBank mRNA | AF538322.1 | - | 7 - 8 | 947 | 86 | |
| AK031345.1 | - | 7 - 8 | 955 | 86 | ||
| AK033395.1 | - | 7 - 8 | 920 | 86 | ||
| AK034217.1 | - | 7 - 8 | 964 | 86 | ||
| AK042892.1 | - | 1 - 2 | 850 | 86 | ||
| AK045758.1 | - | 7 - 8 | 864 | 86 | ||
| BC050065.1 | - | 7 - 8 | 907 | 30016211 | 86 | |
| BC059897.1 | - | 7 - 8 | 860 | 6821433 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dpy19l3
Genomic Map