Assay Details
| Gene Symbol: Mro | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY243876.1 | - | 4 - 5 | 425 | 69 | |
| BY088818.1 | - | 3 - 4 | 306 | 69 | ||
| CK335185.1 | - | 3 - 4 | 291 | 69 | ||
More Information
Set Membership: |
|

Mro
Genomic Map