Assay Details
| Gene Symbol: Kctd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146188.1 | NP_666300.1 | 2 - 3 | 352 | 93 | |
| GenBank mRNA | AK034731.1 | - | 3 - 4 | 424 | 93 | |
| BC031749.1 | - | 2 - 3 | 352 | 4507203 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034731.1, BC031749.1, NM_146188.1 transcripts.
Set Membership: |
|

Kctd15
Genomic Map