| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161369.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161369.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161370.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161370.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_001161371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_009280.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
NM_009280.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800007539.mm.1 |
TC1800001009.mm.1 |
MTA TC |