Assay Details
| Gene Symbol: Cd300lg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160711.1 | NP_001154183.1 | 5 - 6 | 1035 | 70 | |
| NM_001160712.1 | NP_001154184.1 | 5 - 6 | 783 | 70 | ||
| NM_001160713.1 | NP_001154185.1 | 4 - 5 | 903 | 70 | ||
| NM_027987.3 | NP_082263.2 | 4 - 5 | 651 | 70 | ||
| GenBank mRNA | AB243063.1 | - | 5 - 6 | 998 | 70 | |
| AB243064.1 | - | 4 - 5 | 866 | 70 | ||
| AB243065.1 | - | 5 - 6 | 746 | 70 | ||
| AB243066.1 | - | 4 - 5 | 614 | 70 | ||
| AK009375.1 | - | 4 - 5 | 614 | 70 | ||
| AK171982.1 | - | 4 - 5 | 595 | 70 | ||
| AY457055.1 | - | 4 - 5 | 571 | 70 | ||
| BC116906.1 | - | 4 - 5 | 598 | 40126225 | 70 | |
| BC119315.1 | - | 4 - 5 | 598 | 8734064 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009953 | AK171982.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009953 | AK009375.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009953 | AB243065.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009953 | AB243066.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2009953 | AB243063.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 427854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850238 | AK007938.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516363 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458274 | AK047783.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 169025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410412 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 944656 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94726 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171306 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147978 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009953 | AK171982.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009953 | AK009375.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009953 | AB243065.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009953 | AB243066.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2009953 | AB243063.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077107 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077125 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077118 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077144 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077115 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28077147 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171306 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147978 | - | - |
| Whole Mouse Genome G4122A | A_51_P155638 | AK171982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155638 | AK009375.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155638 | AB243065.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155638 | AB243066.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P155638 | AB243063.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193941 | BC119315.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193941 | AK171982.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193941 | BB150844.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193941 | BC116906.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P193941 | AK009375.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147978 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171306 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09566256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09522507 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09522505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09522509 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09483882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28077156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09673442 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09462584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09507476 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09490020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09040213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077130 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077164 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077143 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09526018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077113 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09578654 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09191107 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09483875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09667203 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077138 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077166 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077101 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077133 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09581179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09526017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09716393 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09667195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09462579 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28077117 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10381524 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381515 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381523 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381519 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381525 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381520 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381516 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381521 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381517 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381522 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10381518 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453131_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10381514 | - | - |
| Mouse Genome 430 2.0 Array | 1453131_at | - | - |
| Mouse Genome 430 2.0 Array | 1453131_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52685_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 52685_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160711.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160711.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160712.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160712.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160713.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_001160713.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_027987.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | NM_027987.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100015763.mm.1 | TC1100001677.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 130661_at | - | - |
| Murine Genome U74v2 Set | 166168_PM_f_at | - | - |
| Murine Genome U74v2 Set | 166168_f_at | - | - |
| Murine Genome U74v2 Set | 130661_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453131_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453131_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 450399 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688416 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2574702 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688416 | - | - |

Cd300lg
Genomic Map