Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017437.mm.1 |
NM_207678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017437.mm.1 |
NM_207678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017437.mm.1 |
TC0400002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017434.mm.1 |
NM_207678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017434.mm.1 |
NM_207678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017434.mm.1 |
TC0400002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017435.mm.1 |
NM_207678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017435.mm.1 |
NM_207678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017435.mm.1 |
TC0400002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017436.mm.1 |
NM_207678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017436.mm.1 |
NM_207678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017436.mm.1 |
TC0400002092.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017438.mm.1 |
NM_207678.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017438.mm.1 |
NM_207678.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017438.mm.1 |
TC0400002092.mm.1 |
MTA TC |