Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 3 - 4 | 132 | ||
| GenBank mRNA | AB041605.1 | - | 3 - 4 | 132 | ||
| AF211859.1 | - | 3 - 4 | 132 | |||
| AK007552.1 | - | 3 - 4 | 132 | |||
| AK008585.1 | - | 3 - 4 | 132 | |||
| AK044204.1 | - | 3 - 4 | 132 | |||
| AK048244.1 | - | 3 - 4 | 132 | |||
| AK138938.1 | - | 3 - 4 | 132 | |||
| AK156037.1 | - | 3 - 4 | 487 | 132 | ||
| AK169857.1 | - | 3 - 4 | 480 | 132 | ||
| AY337018.1 | - | 3 - 4 | 132 | |||
| BC023747.1 | - | 3 - 4 | 479 | 132 | ||
| BC132295.1 | - | 3 - 4 | 496 | 40130572 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl2
Genomic Map