Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 5 - 6 | 691 | 108 | |
| GenBank mRNA | AB041605.1 | - | 5 - 6 | 668 | 108 | |
| AY337018.1 | - | 5 - 6 | 674 | 108 | ||
| BC132295.1 | - | 5 - 6 | 683 | 40130572 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl2
Genomic Map