Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 7 - 8 | 900 | 124 | |
| GenBank mRNA | AB041605.1 | - | 7 - 8 | 877 | 124 | |
| AK048244.1 | - | 7 - 8 | 2680 | 124 | ||
| AY337018.1 | - | 7 - 8 | 883 | 124 | ||
| BC023747.1 | - | 7 - 8 | 2665 | 124 | ||
| BC132295.1 | - | 7 - 8 | 892 | 40130572 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccnl2
Genomic Map