Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 9 - 10 | 1151 | 86 | |
| GenBank mRNA | AB041605.1 | - | 9 - 10 | 1128 | 86 | |
| AK048244.1 | - | 9 - 10 | 2931 | 86 | ||
| AY337018.1 | - | 9 - 10 | 1134 | 86 | ||
| BC003773.1 | - | 2 - 3 | 274 | 86 | ||
| BC023747.1 | - | 9 - 10 | 2919 | 86 | ||
| BC083055.1 | - | 2 - 3 | 138 | 86 | ||
| BC132295.1 | - | 9 - 10 | 1143 | 40130572 | 86 | |
| U37351.1 | - | 2 - 3 | 157 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the U37351.1 transcripts.
Set Membership: |
|

Ccnl2
Genomic Map