Assay Details
Gene Symbol: Pop4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025390.4 | NP_079666.1 | 4 - 5 | 558 | 56 | |
GenBank mRNA | AK002855.1 | - | 4 - 5 | 559 | 56 | |
AK003293.2 | - | 4 - 5 | 559 | 56 | ||
AK003908.1 | - | 4 - 5 | 572 | 56 | ||
AK054466.1 | - | 590 | 56 | |||
AK150459.1 | - | 4 - 5 | 565 | 56 | ||
AK152948.1 | - | 4 - 5 | 565 | 56 | ||
BC011465.1 | - | 4 - 5 | 574 | 3966371 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
