Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800020079.mm.1 |
NM_175441.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020079.mm.1 |
NM_175441.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020079.mm.1 |
TC0800002609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020069.mm.1 |
NM_175441.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020069.mm.1 |
NM_175441.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020069.mm.1 |
TC0800002609.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800020070.mm.1 |
NM_175441.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020070.mm.1 |
NM_175441.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800020070.mm.1 |
TC0800002609.mm.1 |
MTA TC |