| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300006587.mm.1 |
NM_011281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300006587.mm.1 |
TC0300000864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300006588.mm.1 |
NM_011281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300006588.mm.1 |
TC0300000864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300006589.mm.1 |
NM_011281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300006589.mm.1 |
TC0300000864.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300006594.mm.1 |
NM_011281.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300006594.mm.1 |
TC0300000864.mm.1 |
MTA TC |