Assay Details
| Gene Symbol: Oaz3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016901.3 | NP_058597.2 | 2 - 3 | 440 | 85 | |
| GenBank mRNA | AB045835.1 | - | 2 - 3 | 365 | 85 | |
| AF175297.1 | - | 2 - 3 | 405 | 85 | ||
| AK006116.1 | - | 2 - 3 | 375 | 85 | ||
| AK006634.1 | - | 2 - 3 | 376 | 85 | ||
| AK076942.1 | - | 2 - 3 | 382 | 85 | ||
| BC119339.1 | - | 2 - 3 | 367 | 85 | ||
| BC119341.1 | - | 2 - 3 | 367 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Oaz3
Genomic Map