Assay Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025506.2 | NP_079782.1 | 1 - 2 | 92 | 146 | |
GenBank mRNA | AK009192.1 | - | 1 - 2 | 94 | 146 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK009192.1, NM_025506.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
