Assay Details
Gene Symbol: Riiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025506.2 | NP_079782.1 | 4 - 5 | 340 | 118 | |
GenBank mRNA | AK009192.1 | - | 4 - 5 | 342 | 118 | |
AK019569.1 | - | 2 - 3 | 150 | 118 | ||
BC107021.1 | - | 3 - 4 | 727 | 40049310 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
