Assay Details
| Gene Symbol: Synpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034012.1 | - | 1 - 2 | 45 | 112 | |
| AK049810.1 | - | 1 - 2 | 105 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK034012.1 transcripts.
Set Membership: |
|

Synpo
Genomic Map