Assay Details
Gene Symbol: Csf1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037859.2 | NP_001032948.2 | 15 - 16 | 2404 | 68 | |
GenBank mRNA | AK004947.1 | - | 14 - 15 | 2214 | 68 | |
AK076215.1 | - | 14 - 15 | 2213 | 68 | ||
AK079247.1 | - | 14 - 15 | 2221 | 68 | ||
AK080326.1 | - | 14 - 15 | 2202 | 68 | ||
AK143545.1 | - | 14 - 15 | 2205 | 68 | ||
AK146069.1 | - | 15 - 16 | 2405 | 68 | ||
AK154128.1 | - | 14 - 15 | 2219 | 68 | ||
AK154556.1 | - | 14 - 15 | 2215 | 68 | ||
AK154653.1 | - | 14 - 15 | 2215 | 68 | ||
AK154699.1 | - | 14 - 15 | 2215 | 68 | ||
AK155488.1 | - | 14 - 15 | 2225 | 68 | ||
AK155640.1 | - | 14 - 15 | 2219 | 68 | ||
AK155654.1 | - | 14 - 15 | 2219 | 68 | ||
AK155858.1 | - | 14 - 15 | 2222 | 68 | ||
AK156550.1 | - | 14 - 15 | 2239 | 68 | ||
AK171386.1 | - | 14 - 15 | 2219 | 68 | ||
BC036343.1 | - | 14 - 15 | 2191 | 4948135 | 68 | |
BC043054.1 | - | 14 - 15 | 2195 | 6418718 | 68 | |
BC050024.1 | - | 14 - 15 | 2302 | 68 | ||
BC066863.1 | - | 15 - 16 | 2313 | 30436119 | 68 | |
X06368.1 | - | 14 - 15 | 2197 | 68 | ||
X68932.1 | - | 14 - 15 | 2107 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
