| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200004015.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004015.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004015.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004018.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004018.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004018.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004012.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004012.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004012.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004013.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004013.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004013.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004016.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004016.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004016.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004021.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004021.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004021.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004017.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004017.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004017.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004019.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004019.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004019.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004020.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004020.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004020.mm.1 |
TC1200000596.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200004022.mm.1 |
NM_028127.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004022.mm.1 |
NM_028127.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200004022.mm.1 |
TC1200000596.mm.1 |
MTA TC |