Assay Details
| Gene Symbol: Psmg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134138.1 | NP_598899.1 | 5 - 6 | 665 | 147 | |
| GenBank mRNA | AB031387.1 | - | 5 - 6 | 665 | 147 | |
| AK077569.1 | - | 5 - 6 | 652 | 147 | ||
| AK167840.1 | - | 5 - 6 | 906 | 147 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792970 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792969 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043009 | AK077569.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043009 | NM_134138.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2043009 | AB031387.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232909 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98003 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 837617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 196144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402218 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 879621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 779362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 657013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 73867 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36976 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P141663 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P104691 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043009 | AK077569.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043009 | NM_134138.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2043009 | AB031387.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792949 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792971 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792969 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792979 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792981 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792951 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792986 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792943 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31792962 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P141663 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09037098 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P104691 | - | - |
| Whole Mouse Genome G4122A | A_51_P338515 | AK167840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338515 | AK077569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338515 | NM_134138.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P338515 | AB031387.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P403352 | AK135578.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P403352 | AK167840.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P403352 | AK077569.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P403352 | BC016606.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P403352 | NM_134138.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101387 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106718 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P104691 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P141663 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13871049 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13871051 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792968 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792975 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792971 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792973 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792974 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31792970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793065 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31792984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31793029 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14031998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14029131 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13957654 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13871049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793024 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792990 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13648839 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13648838 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793050 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793047 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793019 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13994212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792998 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793010 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793012 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793006 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13923042 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14071313 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792992 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793041 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792999 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13979687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13648826 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31793051 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31792982 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10456417 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456418 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456416 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456421 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456422 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456420 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456415 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10456419 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1444264_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448212_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10456414 | - | - |
| Mouse Genome 430 2.0 Array | 1425373_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1444264_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1444264_at | - | - |
| Mouse Genome 430 2.0 Array | 1448212_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1425373_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1448212_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107047_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 107047_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800005194.mm.1 | NM_134138.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800005194.mm.1 | NM_134138.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800005194.mm.1 | TC1800000708.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1800005193.mm.1 | NM_134138.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800005193.mm.1 | NM_134138.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800005193.mm.1 | TC1800000708.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1425373_a_at | - | - |
| Mouse Expression Set 430 | 1448212_PM_at | - | - |
| Mouse Expression Set 430 | 1448212_at | - | - |
| Mouse Expression Set 430 | 1425373_PM_a_at | - | - |
| Murine Genome U74v2 Set | 99129_at | - | - |
| Murine Genome U74v2 Set | 99129_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425373_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448212_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425373_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448212_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448212_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448212_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2060095 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2060095 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002060095 | - | - |
| mouse-6 v1.1 (Illumina) | scl51823.8.1_71-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2060095 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51823.8.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2060095 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3074516 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3153450 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3074516 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3153450 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224609 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl51823.8.1_71-S | - | - |

Psmg2
Genomic Map