Assay Details
| Gene Symbol: Ptpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008977.3 | NP_033003.1 | 9 - 10 | 1238 | 87 | |
| GenBank mRNA | AK014528.1 | - | 9 - 10 | 1205 | 87 | |
| M80739.1 | - | 9 - 10 | 1193 | 87 | ||
| M81477.1 | - | 9 - 10 | 1193 | 87 | ||
| S52655.1 | - | 9 - 10 | 1188 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn2
Genomic Map