Assay Details
Gene Symbol: Tas1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031872.2 | NP_114078.1 | 5 - 6 | 1634 | 63 | |
GenBank mRNA | AB049994.1 | - | 5 - 6 | 1647 | 63 | |
AF311386.1 | - | 5 - 6 | 1615 | 63 | ||
AF337039.1 | - | 5 - 6 | 1634 | 63 | ||
AY032621.1 | - | 5 - 6 | 1615 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
detection of chemical stimulus involved in sensory perception of sweet taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
sensory perception of sweet taste
sensory perception of umami taste
signal transducer activity
G-protein coupled receptor activity
protein binding
taste receptor activity
sweet taste receptor activity
detection of chemical stimulus involved in sensory perception of sweet taste
signal transduction
G-protein coupled receptor signaling pathway
response to stimulus
sensory perception of taste
sensory perception of sweet taste
sensory perception of umami taste
signal transducer activity
G-protein coupled receptor activity
protein binding
taste receptor activity
sweet taste receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 246311 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 538397 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125184 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P423963 | AF337039.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P423963 | AF337039.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441091 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441085 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441086 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441093 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441088 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441105 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441096 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441089 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441106 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441097 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23441101 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05464230 | - | - |
Whole Mouse Genome G4122A | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P423963 | AF337039.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P423963 | NM_031872.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P140882 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121120 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04029781 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04029787 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04029770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440974 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441008 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441157 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441012 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441018 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440988 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441010 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440998 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23440995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23441134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441075 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029787 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441078 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441103 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441112 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029803 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029793 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441106 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03514277 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441108 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04029798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23441091 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10519212 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519215 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519218 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519217 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519216 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519214 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10519213 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1420778_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10519211 | - | - |
Mouse Genome 430 2.0 Array | 1420778_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1420778_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83771_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 83771_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0400034902.mm.1 | TC0400004157.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034902.mm.1 | NM_031872.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034902.mm.1 | NM_031872.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034901.mm.1 | TC0400004157.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400034901.mm.1 | NM_031872.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400034901.mm.1 | NM_031872.2 | Refseq |
Mouse Expression Set 430 | 1420778_PM_at | - | - |
Mouse Expression Set 430 | 1420778_at | - | - |
Mouse Genome 430A 2.0 Array | 1420778_at | - | - |
Mouse Genome 430A 2.0 Array | 1420778_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420778_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1420778_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 639890 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 360136 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000360136 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2593836 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2593836 | - | - |
mouse-6 v1.1 (Illumina) | scl23427.6_143-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 360136 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl23427.6_143-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 360136 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2593836 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2593836 | - | - |
