Assay Details
| Gene Symbol: 2210409D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045360.1 | 1 - 2 | 110 | 76 | ||
| GenBank mRNA | AK008868.1 | - | 1 - 2 | 111 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210409D07Rik| Gene Symbol: 2210409D07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045360.1 | 1 - 2 | 110 | 76 | ||
| GenBank mRNA | AK008868.1 | - | 1 - 2 | 111 | 76 | |
Set Membership: |
|